missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acid and bases.
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents, strong acid and bases.

Specifications
Specifications
Melting Point | 64°C to 67°C |
Quantity | 1 g |
Beilstein | 5262096 |
Solubility Information | Insoluble in water. |
Formula Weight | 184.19 |
Percent Purity | 98% |
Chemical Name or Material | (1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester, 98%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction