missing translation for 'onlineSavingsMsg'
Learn More
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific Chemicals Product Code.: 10120611

(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific Chemicals

Product Code. 10120611
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10120611

Brand: Thermo Scientific Acros 334800250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2964-48-9
Molecular Formula C9H12N2O4
Molecular Weight (g/mol) 212.2
MDL Number MFCD00007359
InChI Key OCYJXSUPZMNXEN-IUCAKERBSA-N
PubChem CID 224171
SMILES C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-]

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 160°C to 166°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 25 g
Specific Rotation + 31
Synonym 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol, 1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol, 1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol, 2-amino-1-4-nitro-phenyl-propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s, threomine, 1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol, 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned, 1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s, l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol
Formula Weight 212.2
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material (1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0318132 Certificate TypeCertificate of Analysis Date28/03/2022 Catalog Number334800250,334801000,334800050
Product Identifier
  • (1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.