missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%, Thermo Scientific™

Product Code. 10257460
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 70749-06-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point 48.0°C to 51.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C16H20N2
MDL Number MFCD00216956
Quantity 250 mg
Specific Rotation Condition − 49.50 (20.00°C c=2,MeOH)
Specific Rotation − 49.50
Synonym 1s,2s-n1,n2-dimethyl-1,2-diphenylethane-1,2-diamine, 1r,2r-n,n'-dimethyl-1,2-diphenyl-1,2-ethylenediamine, 1s,2s-2-methylamino-1,2-diphenylethyl methylamine, 1,2-ethanediamine,n,n'-dimethyl-1,2-diphenyl-, 1s,2s, 1,2-ethanediamine, n,n'-dimethyl-1,2-diphenyl-, 1s,2s, 1s,2s-n,n'-dimethyl-1,2-diphenyl-1,2-ethylenediamine, methyl 1s,2s-2-methylamino-1,2-diphenylethyl amine, 1s,2s-n,n inverted exclamation marka-dimethyl-1,2-diphenyl-1,2-ethylenediamine, 1s,2s-n,n inverted exclamation marka-dimethyl-1,2-diphenylethane-1,2-diamine
InChI Key BTHYVQUNQHWNLS-HOTGVXAUSA-N
SMILES CNC(C1=CC=CC=C1)C(C2=CC=CC=C2)NC
IUPAC Name (1S,2S)-N,N'-dimethyl-1,2-diphenylethane-1,2-diamine
Molecular Weight (g/mol) 240.35
PubChem CID 7019384
Formula Weight 240.35
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material (1S,2S)-(-)-N,N′-Dimethyl-1,2-diphenyl-1,2-ethane diamine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (1S,2S)-(-)-N,N'-Dimethyl-1,2-diphenyl-1,2-ethane diamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'