missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 1X Basal Medium Eagle (BME), liquid
GREENER_CHOICE

1X Basal Medium Eagle (BME), liquid

Brand:  Gibco™ 41010109

Product Code. 11524476

  • 242.00 € / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Basal Medium Eagle (BME) is a widely used synthetic basal medium for supporting the growth of many different mammalian cells. BME was originally developed by Harry Eagle for HeLa cells and mouse fibroblasts, when he discovered the minimum requirements for cell growth in vitro. Gibco™ BME has since been used for other human lines, including WI-38 and MRC-5.

Gibco™ BME contains eight B vitamins, the ten essential amino acids, plus cystine, tyrosine, and glutamine. There are a number of modifications of BME: Minimal Essential Medium, Glasgow's Medium and Dulbecco's Modified Eagle's Medium. This BME formulation contains Earle's salts for use in a CO2 incubator.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law

cGMP Manufacturing and Quality System
Gibco™ BME is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ BME product made in our Grand Island facility (21010-046). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.

BME contains no proteins, lipids, or growth factors. Therefore, BME requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). BME uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ISO 13485 standards, FDA
Basal Medium Eagle (BME), 10 x 500mL, Liquid, Phenol Red Indicator, Low Glucose, 12 Months Shelf Life, Store at 2 to 8°C, Protect From Light, For Mammalian Cell Culture
Basal Medium
No Glutamine, No HEPES, No Sodium Pyruvate
10 x 500 mL
2 to 8°C
Liquid
Phenol Red
Mammalian cell culture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ 1X Basal Medium Eagle (BME), liquid > 10 x 500mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.