missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Thermo Scientific™
106.00 € - 140.00 €
Chemical Identifiers
CAS | 71605-72-6 |
---|---|
Molecular Formula | C7H4N2OS |
Molecular Weight (g/mol) | 164.182 |
MDL Number | MFCD00457949 |
InChI Key | GEFIFDVQYCPLHC-UHFFFAOYSA-N |
Synonym | benzo c 1,2,5 thiadiazole-5-carbaldehyde, 2,1,3-benzothiadiazole-5-carboxaldehyde, benzo 1,2,5 thiadiazole-5-carbaldehyde, 5-formyl-2,1,3-benzothiadiazole, 5-formylbenzo-2,1,3-thiadiazole, benzo 1,2,5 thiadiazol-5-carbaldehyde, benzo 1,2,5 thiadiazole-5-carboxaldehyde |
PubChem CID | 2776290 |
IUPAC Name | 2,1,3-benzothiadiazole-5-carbaldehyde |
SMILES | C1=CC2=NSN=C2C=C1C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10262632
|
Thermo Scientific Maybridge
CC09204CB |
250 mg |
106.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10405312
|
Thermo Scientific Maybridge
CC09204DA |
1 g |
140.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
71605-72-6 | |
164.182 | |
GEFIFDVQYCPLHC-UHFFFAOYSA-N | |
2776290 | |
C1=CC2=NSN=C2C=C1C=O |
C7H4N2OS | |
MFCD00457949 | |
benzo c 1,2,5 thiadiazole-5-carbaldehyde, 2,1,3-benzothiadiazole-5-carboxaldehyde, benzo 1,2,5 thiadiazole-5-carbaldehyde, 5-formyl-2,1,3-benzothiadiazole, 5-formylbenzo-2,1,3-thiadiazole, benzo 1,2,5 thiadiazol-5-carbaldehyde, benzo 1,2,5 thiadiazole-5-carboxaldehyde | |
2,1,3-benzothiadiazole-5-carbaldehyde |
Specifications
71605-72-6 | |
100.0 | |
Amber glass bottle | |
MFCD00457949 | |
GEFIFDVQYCPLHC-UHFFFAOYSA-N | |
2,1,3-benzothiadiazole-5-carbaldehyde | |
2776290 | |
97% |
96.5 | |
97% | |
C7H4N2OS | |
benzo c 1,2,5 thiadiazole-5-carbaldehyde, 2,1,3-benzothiadiazole-5-carboxaldehyde, benzo 1,2,5 thiadiazole-5-carbaldehyde, 5-formyl-2,1,3-benzothiadiazole, 5-formylbenzo-2,1,3-thiadiazole, benzo 1,2,5 thiadiazol-5-carbaldehyde, benzo 1,2,5 thiadiazole-5-carboxaldehyde | |
C1=CC2=NSN=C2C=C1C=O | |
164.182 | |
164.19 | |
2, 1, 3-Benzothiadiazole-5-carbaldehyde |