missing translation for 'onlineSavingsMsg'
Learn More
2,1,3-Benzoxadiazol-4-ylmethanol, Thermo Scientific™ Product Code.: 10632635

2,1,3-Benzoxadiazol-4-ylmethanol, Thermo Scientific™

Product Code. 10632635
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10632635

Brand: Thermo Scientific Maybridge CC09609DE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175609-19-5
Molecular Formula C7H6N2O2
Molecular Weight (g/mol) 150.14
MDL Number MFCD04115376
InChI Key OKDMFOBEJCALFD-UHFFFAOYSA-N
Synonym 4-hydroxymethylbenzofurazan, benzo c 1,2,5 oxadiazol-4-ylmethanol, 4-hydroxymethyl-benzo c 1,2,5 oxadiazole, 2,1,3-benzoxadiazole-4-methanol, 1,2,3-benzoxadiazol-4-ylmethanol, benzo c 1,2,5-oxadiazol-4-ylmethan-1-olShow MoreShow Less
PubChem CID 2795215
IUPAC Name 2,1,3-benzoxadiazol-4-ylmethanol
SMILES OCC1=CC=CC2=NON=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 150.14
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazol-4-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,1,3-Benzoxadiazol-4-ylmethanol, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.