missing translation for 'onlineSavingsMsg'
Learn More
2,1,3-Benzoxadiazole, 97%, Thermo Scientific™ Product Code.: 10514994

2,1,3-Benzoxadiazole, 97%, Thermo Scientific™

Product Code. 10514994
10 g
Click to view available options
Quantity:
1 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 10514994

Brand: Thermo Scientific Maybridge KM06477EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 273-09-6
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.111
InChI Key AWBOSXFRPFZLOP-UHFFFAOYSA-N
Synonym benzofurazan, benzo c 1,2,5 oxadiazole, benzofurazane, 2,1,3-benzoxadiazol, benzo-2,1,3-oxadiazole, 2-oxa-1,3-diaza-2h-isoindene, 3,5-cyclohexadiene-1,2-dione, dioxime, furazan, pubchem22999, acmc-209gwbShow MoreShow Less
PubChem CID 67501
IUPAC Name 2,1,3-benzoxadiazole
SMILES C1=CC2=NON=C2C=C1

Specifications

Assay Percent Range 97%
Quantity 10 g
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzoxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,1,3-Benzoxadiazole, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.