missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Piperazinyl)phenol, 98%, Thermo Scientific Chemicals

Product Code. 11419736
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1011-17-2
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00190246
InChI Key UORNTHBBLYBAJJ-UHFFFAOYSA-N
Synonym 2-piperazin-1-yl phenol, 1-2-hydroxyphenyl piperazine, 2-1-piperazinyl phenol, 2-1-piperazino phenol, o-1-piperazinyl phenol, 2-piperazin-1-yl-phenol, n-2-hydroxyphenyl piperazine, phenol, 2-1-piperazinyl, 1-2-hydroxyphenyl-piperazine, 2-piperazinylphenol
PubChem CID 70530
IUPAC Name 2-piperazin-1-ylphenol
SMILES C1CN(CCN1)C2=CC=CC=C2O

Specifications

Melting Point 122°C to 127°C
Quantity 50 g
UN Number UN3263
Beilstein 10646
Sensitivity Air Sensitive
Formula Weight 178.24
Percent Purity 98%
Chemical Name or Material 2-(1-Piperazinyl)phenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'