missing translation for 'onlineSavingsMsg'
Learn More
[2-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™ Product Code.: 10567874

[2-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™

Product Code. 10567874
10 g
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy

Product Code. 10567874

Brand: Thermo Scientific Maybridge CC34309EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61034-86-4
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD04115123
InChI Key PMFMGYSILUCETA-UHFFFAOYSA-N
Synonym 2-1h-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzyl alcohol, 2-pyrrol-1-yl-phenyl-methanol, 2-pyrrol-1-yl phenyl methanol, 2-1h-pyrrol-1-yl benzylalcohol, pubchem11305, 2-pyrrolylphenyl methan-1-ol, 2-pyrrol-1-yl-benzyl alcohol, 1-2-hydroxymethyl phenyl-1h-pyrroleShow MoreShow Less
PubChem CID 7016493
IUPAC Name (2-pyrrol-1-ylphenyl)methanol
SMILES C1=CC=C(C(=C1)CO)N2C=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 173.21
Percent Purity 97%
Chemical Name or Material [2-(1H-Pyrrol-1-yl)phenyl]methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
[2-(1H-Pyrrol-1-yl)phenyl]methanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.