missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-(1H-Tetrazol-5-yl)phenol, 97%

Product Code. 11362749
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 51449-77-5
Molecular Formula C7H6N4O
Molecular Weight (g/mol) 162.152
MDL Number MFCD09037787
InChI Key SQPPXOJPOBGAAU-UHFFFAOYSA-N
Synonym phenol,2-2h-tetrazol-5-yl, 2-2h-1,2,3,4-tetrazol-5-yl phenol, 5-2-hydroxyphenyl-1h-tetrazole, 2-1h-1,2,3,4-tetrazol-5-yl phenol, acmc-1akh0, 5-o-hydroxyphenyl tetrazole, 2-1h-tetrazole-5-yl phenol, fpuqyvargfdcgz-uhfffaoysa-n, 5-2-hydroxyphenyl-1h-tetrazol, 2-2h-1,2,3,4-tetraazol-5-yl phenol
PubChem CID 42553059
IUPAC Name 6-(1,2-dihydrotetrazol-5-ylidene)cyclohexa-2,4-dien-1-one
SMILES C1=CC(=C2NNN=N2)C(=O)C=C1

Specifications

Melting Point ∽230°C (decomposition)
Quantity 5 g
Formula Weight 162.16
Percent Purity 97%
Chemical Name or Material 2-(1H-Tetrazol-5-yl)phenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'