missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution, Thermo Scientific Chemicals

Product Code. 10422985
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5042-30-8, 7732-18-5
Molecular Formula C2H5F3N2
Molecular Weight (g/mol) 114.07
MDL Number MFCD00007622
InChI Key OPMFFAOEPFATTG-UHFFFAOYSA-N
Synonym 2,2,2-trifluoroethyl hydrazine, 2,2,2-trifluoroethylhydrazine, 70 % in aqueous solution, 2,2,2-trifluoroethyl hydrazine in water, sntljqhjptadtauup@, acmc-20aoks, trifluoroethyl hydrazine, cf3ch2nhnh2, 2,2,2-trifluorethylhydrazine
PubChem CID 78740
IUPAC Name 2,2,2-trifluoroethylhydrazine
SMILES C(C(F)(F)F)NN

Specifications

CAS Min % 65.0, 25.0
CAS Max % 75.0, 35.0
Color Colorless to Yellow
Density 1.2940g/mL
Flash Point 42°C
Packaging Glass bottle
Linear Formula CF3CH2NHNH2
Refractive Index 1.3600 to 1.364
Quantity 5 g
Specific Gravity 1.294
Formula Weight 114.07
Percent Purity ≥65%
Physical Form Liquid
Chemical Name or Material 2, 2, 2-Trifluoroethylhydrazine, 70 wt% aqueous solution
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,2,2-Trifluoroethylhydrazine, 70 wt% aqueous solution
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'