Learn More
2,2,3,3,4,4,4-Heptafluoro-1-butanol, 98%, Thermo Scientific Chemicals
66.83 € - 912.54 €
Chemical Identifiers
CAS | 375-01-9 |
---|---|
Molecular Formula | C4H3F7O |
Molecular Weight (g/mol) | 200.06 |
MDL Number | MFCD00004674 |
InChI Key | WXJFKAZDSQLPBX-UHFFFAOYSA-N |
Synonym | 2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro |
PubChem CID | 9776 |
IUPAC Name | 2,2,3,3,4,4,4-heptafluorobutan-1-ol |
SMILES | OCC(F)(F)C(F)(F)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448613
|
Thermo Scientific Alfa Aesar
A10414.09 |
10 g |
66.83 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444130
|
Thermo Scientific Alfa Aesar
A10414.14 |
25 g |
149.75 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458613
|
Thermo Scientific Alfa Aesar
A10414.18 |
50 g |
253.26 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468613
|
Thermo Scientific Alfa Aesar
A10414.30 |
250 g |
912.54 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a halogenated solvent in electrospray liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is employed as a halogenated solvent in electrospray liquid chromatography.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect from electrostatic charges and keep away from ignition sources.
Chemical Identifiers
375-01-9 | |
200.06 | |
WXJFKAZDSQLPBX-UHFFFAOYSA-N | |
9776 | |
OCC(F)(F)C(F)(F)C(F)(F)F |
C4H3F7O | |
MFCD00004674 | |
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro | |
2,2,3,3,4,4,4-heptafluorobutan-1-ol |
Specifications
375-01-9 | |
95°C to 97°C | |
C4H3F7O | |
1.3 | |
UN1987 | |
2,2,3,3,4,4,4-heptafluoro-1-butanol, 1-butanol, 2,2,3,3,4,4,4-heptafluoro, 1h,1h-heptafluorobutanol-1, 1h,1h-heptafluoro-1-butanol, 1,1-dihydroperfluorobutyl alcohol, 2,2,3,3,4,4,4-heptafluorobutanol, 1,1-dihydroperfluorobutanol, 1,1-h,h-heptafluorobutanol, perfluoro-1,1-dihydrobutanol, butanol, 2,2,3,3,4,4,4-heptafluoro | |
WXJFKAZDSQLPBX-UHFFFAOYSA-N | |
2,2,3,3,4,4,4-heptafluorobutan-1-ol | |
9776 | |
98% |
1.596 | |
25°C (77°F) | |
CF3CF2CF2CH2OH | |
MFCD00004674 | |
1761907 | |
Not miscible or difficult to mix in water. | |
OCC(F)(F)C(F)(F)C(F)(F)F | |
200.06 | |
200.06 | |
2,2,3,3,4,4,4-Heptafluoro-1-butanol |
Safety and Handling
GHS H Statement
H330-H226-H315-H319-H335
Fatal if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.
missing translation for 'einecsNumber' : 206-782-1
missing translation for 'rtecsNumber' : EL4130000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only