missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid, Thermo Scientific™

Product Code. 10653635
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 257876-07-6
Molecular Formula C10H5Cl2NO2S
Molecular Weight (g/mol) 274.115
InChI Key ANTJCNIMRNBFMJ-UHFFFAOYSA-N
Synonym 2-2,3-dichlorophenyl-1,3-thiazole-4-carboxylic acid, 2-2,3-dichloro-phenyl-thiazole-4-carboxylic acid, 2-2,3-dichlorophenyl thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-2,3-dichlorophenyl
PubChem CID 2743987
IUPAC Name 2-(2,3-dichlorophenyl)-1,3-thiazole-4-carboxylic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C2=NC(=CS2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 274.13
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dichlorophenyl)-1, 3-thiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'