missing translation for 'onlineSavingsMsg'
Learn More
2,2,4,4-Tetrafluoro-1,3-dithietane, 97%, Thermo Scientific Chemicals Product Code.: 11447244

2,2,4,4-Tetrafluoro-1,3-dithietane, 97%, Thermo Scientific Chemicals

Product Code. 11447244
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11447244

Brand: Thermo Scientific Alfa Aesar B21620.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1717-50-6
Molecular Formula C2F4S2
Molecular Weight (g/mol) 164.14
MDL Number MFCD00053538
InChI Key MPOCEMBRPHOCMK-UHFFFAOYSA-N
Synonym tetrafluoro-1,3-dithietane, pubchem3989, acmc-1budt, 1, 2,2,4,4-tetrafluoro, 2,4,4-tetrafluoro-1,3-dithietane, 1,3-dithietane,2,2,4,4-tetrafluoro, 2,2,4,4-tetrafluoro-1,3-dithietaneShow MoreShow Less
PubChem CID 74392
IUPAC Name 2,2,4,4-tetrafluoro-1,3-dithietane
SMILES FC1(F)SC(F)(F)S1

Specifications

Density 1.6
Boiling Point 47°C to 49°C
Flash Point 36°C (96°F)
Odor Unpleasant
Refractive Index 1.393
Quantity 50 g
UN Number UN1993
Formula Weight 164.14
Percent Purity 97%
Chemical Name or Material 2,2,4,4-Tetrafluoro-1,3-dithietane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10218784 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21620.09,B21620.18
Lot NumberG17T014 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21620.09,B21620.18
Lot Number10139787 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21620.09,B21620.18
Lot Number10211173 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21620.09,B21620.18
Lot Number10224717 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB21620.09,B21620.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2,4,4-Tetrafluoro-1,3-dithietane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.