Learn More
2,2,4,4-Tetramethylpentane, 98%, Thermo Scientific Chemicals
88.40 € - 290.00 €
Chemical Identifiers
CAS | 1070-87-7 |
---|---|
Molecular Formula | C9H20 |
Molecular Weight (g/mol) | 128.26 |
MDL Number | MFCD00060873 |
InChI Key | GUMULFRCHLJNDY-UHFFFAOYSA-N |
PubChem CID | 14058 |
IUPAC Name | 2,2,4,4-tetramethylpentane |
SMILES | CC(C)(C)CC(C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490197
|
Thermo Scientific Alfa Aesar
B21642.09 |
10 g |
88.40 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400207
|
Thermo Scientific Alfa Aesar
B21642.18 |
50 g |
290.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1070-87-7 | |
128.26 | |
GUMULFRCHLJNDY-UHFFFAOYSA-N | |
2,2,4,4-tetramethylpentane |
C9H20 | |
MFCD00060873 | |
14058 | |
CC(C)(C)CC(C)(C)C |
Specifications
1070-87-7 | |
121°C | |
Petroleum-like | |
(CH3)3CCH2C(CH3)3 | |
UN3295 | |
GUMULFRCHLJNDY-UHFFFAOYSA-N | |
2,2,4,4-tetramethylpentane | |
14058 | |
98% |
0.72 | |
6°C (43°F) | |
C9H20 | |
MFCD00060873 | |
di-tert-butylmethane, pentane, 2,2,4,4-tetramethyl, unii-7js6eal3ee, 7js6eal3ee, pentane, 2,2,4,4-tetramethyl-8ci 9ci, 2,4,4-tetramethylpentane, acmc-2098nd, pentane,2,4,4-tetramethyl, 2,2,4,4,-tetramethylpentane, pentane,2,2,4,4-tetramethyl | |
CC(C)(C)CC(C)(C)C | |
128.26 | |
128.26 | |
2,2,4,4-Tetramethylpentane |
Safety and Handling
GHS H Statement
H225
Highly flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
H225-H304
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only