missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ULTRA RESI-ANALYZED, J.T. Baker™
Click to view available options
Quantity:
4 L
Packaging:
Glass Container
Specifications
Specifications
CAS | 540-84-1 |
Flash Point | −12°C |
Packaging | Glass Container |
Molecular Formula | C8H18 |
Quantity | 4 L |
Specific Gravity | 1L=0.69kg |
Synonym | isooctane, isobutyltrimethylmethane, pentane, 2,2,4-trimethyl, iso-octane, 2,4,4-trimethylpentane, unii-qab8f5669o, ch3 2chch2c ch3 3, 2, 2, 4-trimethylpentane, i-octane, isobutyltrimethylethane |
InChI Key | NHTMVDHEPJAVLT-UHFFFAOYSA-N |
SMILES | CC(C)CC(C)(C)C |
IUPAC Name | 2,2,4-trimethylpentane |
Show More |
Safety and Handling
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'