Learn More
2,2,6-Trimethyl-1,3-dioxin-4-one, cont. up to ca 6% acetone, Thermo Scientific Chemicals
30.65 € - 267.33 €
Chemical Identifiers
CAS | 5394-63-8 |
---|---|
Molecular Formula | C7H10O3 |
Molecular Weight (g/mol) | 142.154 |
MDL Number | MFCD00040468 |
InChI Key | XFRBXZCBOYNMJP-UHFFFAOYSA-N |
Synonym | 2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb |
PubChem CID | 79368 |
IUPAC Name | 2,2,6-trimethyl-1,3-dioxin-4-one |
SMILES | CC1=CC(=O)OC(O1)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432387
|
Thermo Scientific Alfa Aesar
A14254.14 |
25 g |
30.65 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442387
|
Thermo Scientific Alfa Aesar
A14254.22 |
100 g |
87.74 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452387
|
Thermo Scientific Alfa Aesar
A14254.36 |
500 g |
267.33 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,6-Trimethyl-1,3-dioxin-4-one is used in the synthesis of dihydropyrimininones as picomolar sodium iodide symporter inhibitors. It is also used in the synthesis of resorcinol amide Hsp90 Inhibitor AT13387. It is used in the synthesis of acetylketene by flash pyrolysis.
Solubility
Soluble in Chloroform, Methanol. Not miscible in water.
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Chemical Identifiers
5394-63-8 | |
142.154 | |
XFRBXZCBOYNMJP-UHFFFAOYSA-N | |
79368 | |
CC1=CC(=O)OC(O1)(C)C |
C7H10O3 | |
MFCD00040468 | |
2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb | |
2,2,6-trimethyl-1,3-dioxin-4-one |
Specifications
5394-63-8 | |
1.094 | |
14°C (57°F) | |
1.461 | |
UN1993 | |
2,2,6-trimethyl-4h-1,3-dioxin-4-one, 2,2,4-trimethyl-6-keto-1,3-dioxin, diketene acetone adduct, 4h-1,3-dioxin-4-one, 2,2,6-trimethyl, 2,2,6-trimethyl-1,3-dioxen-4-one, 2,2,6-trimethyl-1,3-dioxine-4-one, 2,2,6-trimethyl-1,3 dioxin-4-one, pubchem20364, 2,2,6-trimethyl-4h-1,3-dioxin-4-on, acmc-1akcb | |
XFRBXZCBOYNMJP-UHFFFAOYSA-N | |
2,2,6-trimethyl-1,3-dioxin-4-one | |
79368 | |
94% | |
2,2,6-Trimethyl-1,3-dioxin-4-one |
7°C | |
65°C to 67°C (2 mmHg) | |
C7H10O3 | |
MFCD00040468 | |
2408 | |
Soluble in Chloroform,Methanol. Not miscible in water. | |
CC1=CC(=O)OC(O1)(C)C | |
142.154 | |
142.15 | |
cont. up to ≈6% acetone |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
H225-H319-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'einecsNumber' : 226-403-3
missing translation for 'tsca' : No
Recommended Storage : Keep cold
- 2,2,6-Trimethyl-1,3-dioxin-4-one, contains up to 6% acetone
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : 2,2,6-Trimethyl-4H-1,3-dioxin-4-one
RUO – Research Use Only