missing translation for 'onlineSavingsMsg'
Learn More
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals Product Code.: 10438172

(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals

Product Code. 10438172
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10438172

Brand: Thermo Scientific Acros 368640050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98327-87-8
Molecular Formula C44H32P2
Molecular Weight (g/mol) 622.69
MDL Number MFCD00010805
InChI Key MUALRAIOVNYAIW-UHFFFAOYSA-N
Synonym binap, r-+-2,2'-bis diphenylphosphino-1,1'-binaphthyl, 2,2'-bis diphenylphosphino-1,1'-binaphthyl, s-binap, s---binap, s---2,2'-bis diphenylphosphino-1,1'-binaphthyl, r-+-binap, r-binap, rac-binap, 2,2'-bis diphenylphosphino-1,1'-binaphthaleneShow MoreShow Less
PubChem CID 634876
IUPAC Name [1-(2-diphenylphosphanylnaphthalen-1-yl)naphthalen-2-yl]-diphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=C(C2=CC=CC=C2C=C1)C1=C(C=CC2=CC=CC=C12)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 280.0°C to 285.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ((C6H5)2PC10H2)2
Quantity 5 g
Merck Index 15,1227
Formula Weight 622.69
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (±)-2, 2'-Bis(diphenylphosphino)-1, 1'-binaphthyl
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458983 Certificate TypeCertificate of Analysis Date16/02/2024 Catalog Number368640010,368640250,368640050,368641000
Lot NumberA0432284 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number368640010,368640250,368640050,368641000
Lot NumberA0420400 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number368640010,368640250,368640050,368641000
Lot NumberA0413640 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number368640010,368640250,368640050,368641000
Lot NumberA0419285 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number368640010,368640250,368640050,368641000
compliance-icons
Product Identifier
  • (#1)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.