missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Bithiophene, 97%, Thermo Scientific Chemicals

Product Code. 10117420
Click to view available options
Quantity:
2.5 g
20 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 492-97-7
Molecular Formula C8H6S2
Molecular Weight (g/mol) 166.27
MDL Number MFCD00005414
InChI Key OHZAHWOAMVVGEL-UHFFFAOYSA-N
Synonym 2,2'-bithiophene, 2,2'-bithienyl, dithienyl, bithiophene, 2,2'-dithienyl, 2,2' bithiophenyl, 2,2-bithiophene, 2,2'-dithiophene, alpha-bithiophene, 2-2-thienyl thiophene
PubChem CID 68120
ChEBI CHEBI:36821
IUPAC Name 2-thiophen-2-ylthiophene
SMILES C1=CSC(=C1)C2=CC=CS2

Spécification

CAS Min % 96.0
CAS Max % 100.0
Melting Point 30.0°C to 35.0°C
Boiling Point 260.0°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 2.5 g
Beilstein 19, 32
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol
Formula Weight 166.27
Percent Purity 97%
Chemical Name or Material 2, 2'-Bithiophene, 97%
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 2,2'-Bithiophene
Mention d'avertissement
  • Warning
Catégorie de danger
  • Acute toxicity Category 4
Mention de danger
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'