missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Bromoethoxy)anisole, 97+%, Thermo Scientific™
44.50 € - 552.75 €
Chemical Identifiers
CAS | 4463-59-6 |
---|---|
Molecular Formula | C9H11BrO2 |
Molecular Weight (g/mol) | 231.089 |
MDL Number | MFCD00017892 |
InChI Key | PBRPKYRJVDJZTF-UHFFFAOYSA-N |
Synonym | 2-2-bromoethoxy anisole, 1-2-bromoethoxy-2-methoxybenzene, 2-2-bromoethoxy-1-methoxybenzene, 2-bromoethyl 2-methoxyphenyl ether, 2-2-methoxyphenoxy ethylbromide, pubchem3133, 2-methoxyphenoxy ethylbromide, ksc495i5p, akos bc-2659, timtec-bb sbb008602 |
PubChem CID | 1799099 |
IUPAC Name | 1-(2-bromoethoxy)-2-methoxybenzene |
SMILES | COC1=CC=CC=C1OCCBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15449077
|
Thermo Scientific Alfa Aesar
H52382.MD |
250 mg | N/A | ||||||
15429077
|
Thermo Scientific Alfa Aesar
H52382.03 |
1 g | N/A | ||||||
15439077
|
Thermo Scientific Alfa Aesar
H52382.06 |
5 g |
552.75 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
4463-59-6 | |
231.089 | |
PBRPKYRJVDJZTF-UHFFFAOYSA-N | |
1799099 | |
COC1=CC=CC=C1OCCBr |
C9H11BrO2 | |
MFCD00017892 | |
2-2-bromoethoxy anisole, 1-2-bromoethoxy-2-methoxybenzene, 2-2-bromoethoxy-1-methoxybenzene, 2-bromoethyl 2-methoxyphenyl ether, 2-2-methoxyphenoxy ethylbromide, pubchem3133, 2-methoxyphenoxy ethylbromide, ksc495i5p, akos bc-2659, timtec-bb sbb008602 | |
1-(2-bromoethoxy)-2-methoxybenzene |
Specifications
4463-59-6 | |
MFCD00017892 | |
PBRPKYRJVDJZTF-UHFFFAOYSA-N | |
1-(2-bromoethoxy)-2-methoxybenzene | |
1799099 | |
≥97% |
C9H11BrO2 | |
2-2-bromoethoxy anisole, 1-2-bromoethoxy-2-methoxybenzene, 2-2-bromoethoxy-1-methoxybenzene, 2-bromoethyl 2-methoxyphenyl ether, 2-2-methoxyphenoxy ethylbromide, pubchem3133, 2-methoxyphenoxy ethylbromide, ksc495i5p, akos bc-2659, timtec-bb sbb008602 | |
COC1=CC=CC=C1OCCBr | |
231.089 | |
231.09 | |
2-(2-Bromoethoxy)anisole |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title