missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-Bromophenyl)ethylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
Description
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
2-(2-Bromophenyl)ethylamine is used as a pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
Density | 1.396 |
Boiling Point | 253°C |
Refractive Index | 1.577 |
Quantity | 10 g |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Formula Weight | 200.08 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2-Bromophenyl)ethylamine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction