missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(2-chlorophenoxy)ethanimidamide hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Linear Formula | ClC6H4OCH2C(=NH)NH2·HCl |
Quantity | 1 g |
IUPAC Name | hydrogen 2-(2-chlorophenoxy)ethanimidamide chloride |
Formula Weight | 221.09 |
Percent Purity | 97% |
Chemical Name or Material | 2-(2-Chlorophenoxy)ethanimidamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'