missing translation for 'onlineSavingsMsg'
Learn More

2,2-Di-n-propylacetyl chloride, 98%, Thermo Scientific Chemicals

67.64 € - 231.15 €

Chemical Identifiers

CAS 2936-08-5
Molecular Formula C8H15ClO
Molecular Weight (g/mol) 162.66
MDL Number MFCD00051446
InChI Key PITHYUDHKJKJNQ-UHFFFAOYSA-N
Synonym 2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
PubChem CID 76240
IUPAC Name 2-propylpentanoyl chloride
SMILES CCCC(CCC)C(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11362746
View Documents
Thermo Scientific Alfa Aesar
L08852.06
5 g
67.64 €
5g
Please to purchase this item. Need a web account? Register with us today!
11372746
View Documents
Thermo Scientific Alfa Aesar
L08852.14
25 g
231.15 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2-Di-n-propylacetyl chloride is used to produce 1,2-bis-(2-propyl-valerylamino)-ethan.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible materials are Oxidizing agents and bases. Store away from moisture and strong bases.
Specifications

Chemical Identifiers

2936-08-5
162.66
PITHYUDHKJKJNQ-UHFFFAOYSA-N
76240
CCCC(CCC)C(=O)Cl
C8H15ClO
MFCD00051446
2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
2-propylpentanoyl chloride

Specifications

2936-08-5
68°C to 70°C (10 mmHg)
MFCD00051446
1098856
2-propylvaleryl chloride, valproyl chloride, 2,2-di-n-propylacetyl chloride, dipropylacetyl chloride, 2-propylvaleroyl chloride, 2-n-propyl-n-valeroyl chloride, valproic acid chloride, di-n-propylacetyl chloride, acmc-2097cd, 2-n-propylvaleroyl chloride
PITHYUDHKJKJNQ-UHFFFAOYSA-N
2-propylpentanoyl chloride
76240
98%
0.95
C8H15ClO
UN3265
Moisture sensitive
Reacts with water.
CCCC(CCC)C(=O)Cl
162.66
162.66
2,2-Di-n-propylacetyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 220-914-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only