missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Diaminodiphenyl disulfide, 97%, Thermo Scientific Chemicals

Product Code. 15492158
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,2'-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′-Diaminodiphenyl disulfide, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used as a primary and secondary intermediate.

Solubility
Soluble in hot methanol- almost transparent.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1141-88-4
Molecular Formula C12H12N2S2
Molecular Weight (g/mol) 248.36
MDL Number MFCD00007703
InChI Key YYYOQURZQWIILK-UHFFFAOYSA-N
Synonym 2,2'-dithiodianiline, benzenamine, 2,2'-dithiobis, intramine, 2,2'-disulfanediyldianiline, 2-aminophenyl disulfide, 2,2'-diaminodiphenyldisulfide, 2,2'-diaminodiphenyl disulfide, 2-2-aminophenyl disulfanyl aniline, aniline, 2,2'-dithiodi, bis o-aminophenyl disulfide
PubChem CID 14358
IUPAC Name 2-[(2-aminophenyl)disulfanyl]aniline
SMILES NC1=CC=CC=C1SSC1=CC=CC=C1N

Specifications

Melting Point 91°C to 92°C
Quantity 5 g
Beilstein 914032
Solubility Information Soluble in hot methanol- almost transparent.
Formula Weight 248.36
Percent Purity 97%
Chemical Name or Material 2,2'-Diaminodiphenyl disulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'