missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Diethoxyethanethioamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 10 g |
Synonym | 2,2-diethoxy-ethanethioamide, diethoxythioacetamide, 2,2-diethoxythioacetamide, 2,2,-diethoxyethanethioamide, ethanethioamide, 2,2-diethoxy, ethanethioamide,2,2-diethoxy, 2,2-bis ethyloxy ethanethioamide, 1-amino-2,2-diethoxyethane-1-thione |
Formula Weight | 163.24 |
Percent Purity | 97% |
Chemical Name or Material | 2, 2-Diethoxyethanethioamide |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2,2-Diethoxyethanethioamide, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction