missing translation for 'onlineSavingsMsg'
Learn More
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals Product Code.: 15418295

(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 15418295
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15418295

Brand: Thermo Scientific Alfa Aesar H33055.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 128073-33-6
Molecular Formula C5H6F2O2
Molecular Weight (g/mol) 136.098
MDL Number MFCD07777163
InChI Key HLFLYOQLHYYNLT-UHFFFAOYSA-N
Synonym 2,2-difluoro-1-methylcyclopropanecarboxylic acid, -2,2-difluoro-1-methylcyclopropanecarboxylic acid, +/--2,2-difluoro-1-methylcyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane carboxylic acid, 2,2-difluoro-1-methyl-1-cyclopropanecarboxylic acid, 2,2-difluoro-1-methyl-cyclopropanecarboxylic acid, 2,2-difluoro-1-methylcyclopropane-carboxylic acid, -2,2-difluoro-1-methylcyclopropane-carboxylicacid, 2,2-difluoro-1-methyl cyclopropane carboxylic acid, +/--2,2-difluoro-1-methylcyclopropane-carboxylicacidShow MoreShow Less
PubChem CID 14695229
IUPAC Name 2,2-difluoro-1-methylcyclopropane-1-carboxylic acid
SMILES CC1(CC1(F)F)C(=O)O

Specifications

Melting Point 62°C to 64°C
Quantity 250 mg
Formula Weight 136.1
Percent Purity 97%
Chemical Name or Material (±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253207 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog NumberH33055.MD,H33055.03
Lot Number10225552 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33055.MD,H33055.03
Lot Number10211274 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33055.MD,H33055.03
Lot Number10168970 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH33055.MD,H33055.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(±)-2,2-Difluoro-1-methylcyclopropanecarboxylic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.