missing translation for 'onlineSavingsMsg'
Learn More
2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™ Product Code.: 15456236

2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™

Product Code. 15456236
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15456236

Brand: Thermo Scientific Alfa Aesar H61062.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Ticagrelor intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85160-83-4
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD11048438
InChI Key KGWYDVZSGRHPFK-UHFFFAOYSA-N
Synonym 2,2-dimethyl-7-nitro-2h-benzo b 1,4 oxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzoxazin-3 4h-one, 2,2-dimethyl-7-nitro-2h-1,4-benzooxazine-3 4h-oneShow MoreShow Less
PubChem CID 12793543
IUPAC Name 2,2-dimethyl-7-nitro-4H-1,4-benzoxazin-3-one
SMILES CC1(C)OC2=C(NC1=O)C=CC(=C2)[N+]([O-])=O

Specifications

Quantity 1 g
Formula Weight 222.2
Percent Purity 97%
Chemical Name or Material 2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberH22Z021 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog NumberH61062.03,H61062.MD
Lot NumberH22Z021 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2-Dimethyl-7-nitro-2H-1,4-benzoxazin-3(4H)-one, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.