missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethyl-N-(4-pyridinyl)propanamide, 97%, Thermo Scientific™

Product Code. 10601113
10 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 70298-89-4
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.235
MDL Number MFCD00996248
InChI Key JCMMVFHXRDNILC-UHFFFAOYSA-N
Synonym 2,2-dimethyl-n-pyridin-4-yl-propionamide, n-pyridin-4-yl pivalamide, 2,2-dimethyl-n-pyridin-4-yl propanamide, 2,2-dimethyl-n-4-pyridinyl propanamide, n-4-pyridyl pivalamide, 2,2-dimethyl-n-4-pyridyl propanamide, 4-2,2,2-trimethylacetamido pyridine, 2,2-dimethyl-n-4-pyridyl propionamide, 4-pivaloylaminopyridine, maybridge3_004407
PubChem CID 427059
IUPAC Name 2,2-dimethyl-N-pyridin-4-ylpropanamide
SMILES CC(C)(C)C(=O)NC1=CC=NC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethyl-N-(4-pyridinyl)propanamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2-Dimethyl-N-(4-pyridinyl)propanamide, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.