missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylcyclohexanone, Thermo Scientific Chemicals

253.00 € - 870.00 €

Chemical Identifiers

CAS 1193-47-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.2
InChI Key KNSPBSQWRKKAPI-UHFFFAOYSA-N
Synonym 2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
PubChem CID 136929
IUPAC Name 2,2-dimethylcyclohexan-1-one
SMILES CC1(CCCCC1=O)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10051074
View Documents
Thermo Scientific Acros
426500010
1 g Glass bottle
253.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10376254
View Documents
Thermo Scientific Acros
426500050
5 g Glass bottle
870.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1193-47-1
126.2
2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
2,2-dimethylcyclohexan-1-one
C8H14O
KNSPBSQWRKKAPI-UHFFFAOYSA-N
136929
CC1(CCCCC1=O)C

Specifications

1193-47-1
Colorless to Yellow
169.0°C to 170.0°C (0.8 mmHg)
Authentic
(CH3)2C6H8(=O)
0.912
KNSPBSQWRKKAPI-UHFFFAOYSA-N
2,2-dimethylcyclohexan-1-one
136929
≥94%
2, 2-Dimethylcyclohexanone
-20.0°C
0.9120g/mL
50°C
C8H14O
1.4470 to 1.4490 (20°C, 589nm)
2,2-dimethylcyclohexanone, cyclohexanone, 2,2-dimethyl, dimethylcyclohexanone, cyclohexanone, dimethyl, 6,6-dimethylcyclohexanone, cyclohexanone,2-dimethyl, acmc-20a0u6, 2,2-dimethyl-cyclohexanone, cyclohexanone,2,2-dimethyl, 2,2-dimethyl-1-cyclohexanone
CC1(CCCCC1=O)C
126.2
126.2
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

Product Identifier
  • 2,2-Dimethylcyclohexanone

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only