missing translation for 'onlineSavingsMsg'
Learn More

2,2-Dimethylpropanimidamide hydrochloride, 97%, Thermo Scientific™

Product Code. p-2696648
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10193124

Brand: Thermo Scientific Maybridge RF01501DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 18202-73-8
Molecular Formula C5H13ClN2
Molecular Weight (g/mol) 136.623
MDL Number MFCD00051988
InChI Key ARDGQYVTLGUJII-UHFFFAOYSA-N
Synonym 2,2-dimethylpropanimidamide hydrochloride, pivalamidine hydrochloride, pivalimidamide hydrochloride, tert-butylcarbamidine hydrochloride, 2,2-dimethylpropionamidine hydrochloride, 2,2-dimethyl-propionamidine hydrochloride, 2,2-dimethylpropionamidine hcl, t-butylcarbamidine hydrochloride, 2,2-dimethylpropanamidine hydrochloride, 2,2,2-trimethylacetamidine hydrochloride
PubChem CID 2781880
IUPAC Name 2,2-dimethylpropanimidamide;hydrochloride
SMILES CC(C)(C)C(=N)N.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula (CH3)3CC(=NH)NH2·HCl
Quantity 1 g
Formula Weight 136.62
Percent Purity 97%
Chemical Name or Material 2, 2-Dimethylpropanimidamide hydrochloride
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Specific target organ toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Pivalamidine hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.