missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Dithiosalicylic acid, 96%, Thermo Scientific Chemicals

Product Code. 11396846
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-80-2
Molecular Formula C14H10O4S2
Molecular Weight (g/mol) 306.35
MDL Number MFCD00002465
InChI Key LBEMXJWGHIEXRA-UHFFFAOYSA-N
Synonym 2,2'-dithiodibenzoic acid, 2,2'-dithiosalicylic acid, benzoic acid, 2,2'-dithiobis, 2-2-carboxyphenyl disulfanyl benzoic acid, bis 2-carboxyphenyl disulfide, bis o-carboxylphenyl disulfide, 2,2-dithiosalicylic acid, 2,2'-dithiobisbenzoic acid, unii-58mgb5279d, 2,2'-dithiodi benzoic acid
PubChem CID 8409
IUPAC Name 2-[(2-carboxyphenyl)disulfanyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)SSC2=CC=CC=C2C(=O)O

Specifications

Melting Point 287°C to 289°C
Odor Odorless
Quantity 500 g
Beilstein 2221810
Formula Weight 306.36
Percent Purity 96%
Chemical Name or Material 2,2'-Dithiosalicylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'