missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethylamine, 97%, Thermo Scientific™

Product Code. 10737485
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 165115-15-1
Molecular Formula C6H10N2S
Molecular Weight (g/mol) 142.22
MDL Number MFCD03011601
InChI Key GLEZGSKBHYTSAD-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl ethanamine, 2-2-methylthiazol-4-yl ethanamine, 2-2-methyl-1,3-thiazol-4-yl ethylamine, 4-thiazoleethanamine, 2-methyl, 2-2-methyl-1,3-thiazol-4-yl ethan-1-amine, 4-thiazoleethanamine,2-methyl, 2-2-methyl-1,3-thiazol-4-yl-1-ethanamine
PubChem CID 7536518
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)ethanamine
SMILES CC1=NC(CCN)=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 142.23
Percent Purity 97%
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.