missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Methyl-4-thiazolyl)phenol, 97%, Thermo Scientific™

68.50 € - 220.00 €

Chemical Identifiers

CAS 160241-65-6
Molecular Formula C10H9NOS
Molecular Weight (g/mol) 191.248
MDL Number MFCD00665949
InChI Key LYLPLPWQCLBGHW-UHFFFAOYSA-N
Synonym 2-2-methylthiazol-4-yl phenol, 2-2-methyl-1,3-thiazol-4-yl phenol, 2-2-methyl-4-thiazolyl phenol, acmc-1c1by, phenol,2-2-methyl-4-thiazolyl, 2-2-methyl-thiazol-4-yl-phenol
PubChem CID 18801453
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)phenol
SMILES CC1=NC(=CS1)C2=CC=CC=C2O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11355459
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50431.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11365459
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H50431.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

160241-65-6
191.248
LYLPLPWQCLBGHW-UHFFFAOYSA-N
18801453
CC1=NC(=CS1)C2=CC=CC=C2O
C10H9NOS
MFCD00665949
2-2-methylthiazol-4-yl phenol, 2-2-methyl-1,3-thiazol-4-yl phenol, 2-2-methyl-4-thiazolyl phenol, acmc-1c1by, phenol,2-2-methyl-4-thiazolyl, 2-2-methyl-thiazol-4-yl-phenol
2-(2-methyl-1,3-thiazol-4-yl)phenol

Specifications

160241-65-6
C10H9NOS
2-2-methylthiazol-4-yl phenol, 2-2-methyl-1,3-thiazol-4-yl phenol, 2-2-methyl-4-thiazolyl phenol, acmc-1c1by, phenol,2-2-methyl-4-thiazolyl, 2-2-methyl-thiazol-4-yl-phenol
CC1=NC(=CS1)C2=CC=CC=C2O
191.248
191.25
2-(2-Methyl-4-thiazolyl)phenol
61°C to 64°C
MFCD00665949
LYLPLPWQCLBGHW-UHFFFAOYSA-N
2-(2-methyl-1,3-thiazol-4-yl)phenol
18801453
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications