Learn More
2-(2-Naphthyl)acetyl chloride, 97%, Thermo Scientific™
53.10 € - 278.00 €
Chemical Identifiers
CAS | 37859-25-9 |
---|---|
Molecular Formula | C12H9ClO |
Molecular Weight (g/mol) | 204.653 |
MDL Number | MFCD02180524 |
InChI Key | QEJGMKHQXSZCOS-UHFFFAOYSA-N |
Synonym | 2-2-naphthyl acetyl chloride, naphthalen-2-yl-acetyl chloride, 2-naphthylacetyl chloride, 2-naphthalen-2-yl acetyl chloride, 2-naphthaleneacetyl chloride, naphth-2-yl acetyl chloride, zlchem 122, pubchem15724, 2-naphthylacetyl chlotide, beta-naphthylacetyl chloride |
PubChem CID | 588115 |
IUPAC Name | 2-naphthalen-2-ylacetyl chloride |
SMILES | C1=CC=C2C=C(C=CC2=C1)CC(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10573232
|
Thermo Scientific Maybridge
PD00828DA |
1 g | Amber Glass Bottle |
53.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10643603
|
Thermo Scientific Maybridge
PD00828EA |
10 g | Amber Glass Bottle |
278.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
37859-25-9 | |
204.653 | |
QEJGMKHQXSZCOS-UHFFFAOYSA-N | |
588115 | |
C1=CC=C2C=C(C=CC2=C1)CC(=O)Cl |
C12H9ClO | |
MFCD02180524 | |
2-2-naphthyl acetyl chloride, naphthalen-2-yl-acetyl chloride, 2-naphthylacetyl chloride, 2-naphthalen-2-yl acetyl chloride, 2-naphthaleneacetyl chloride, naphth-2-yl acetyl chloride, zlchem 122, pubchem15724, 2-naphthylacetyl chlotide, beta-naphthylacetyl chloride | |
2-naphthalen-2-ylacetyl chloride |
Specifications
37859-25-9 | |
97% | |
MFCD02180524 | |
QEJGMKHQXSZCOS-UHFFFAOYSA-N | |
2-naphthalen-2-ylacetyl chloride | |
588115 | |
97% |
97.0 | |
C12H9ClO | |
2-2-naphthyl acetyl chloride, naphthalen-2-yl-acetyl chloride, 2-naphthylacetyl chloride, 2-naphthalen-2-yl acetyl chloride, 2-naphthaleneacetyl chloride, naphth-2-yl acetyl chloride, zlchem 122, pubchem15724, 2-naphthylacetyl chlotide, beta-naphthylacetyl chloride | |
C1=CC=C2C=C(C=CC2=C1)CC(=O)Cl | |
204.653 | |
204.66 | |
2-(2-Naphthyl)acetyl chloride |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger