missing translation for 'onlineSavingsMsg'
Learn More

2',3',4',5',6'-Pentamethyl-2-phenylacetophenone, 95%, Thermo Scientific™

Product Code. 15428775
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147834-57-9
Molecular Formula C19H22O
Molecular Weight (g/mol) 266.384
MDL Number MFCD00052403
InChI Key CFSAHKOFVAHZDH-UHFFFAOYSA-N
Synonym 1-2,3,4,5,6-pentamethylphenyl-2-phenylethan-1-one, 1-2,3,4,5,6-pentamethylphenyl-2-phenylethanone, ethanone,1-pentamethylphenyl-2-phenyl-9ci, 1-pentamethylphenyl-2-phenylethanone, acmc-20n5au, maybridge1_002825, 1-pentamethylphenyl-2-phenylethan-1-one, 2',3',4',5',6'-pentamethyl-2-phenylacetophenone
PubChem CID 2776715
IUPAC Name 1-(2,3,4,5,6-pentamethylphenyl)-2-phenylethanone
SMILES CC1=C(C(=C(C(=C1C)C)C(=O)CC2=CC=CC=C2)C)C

Specifications

Melting Point 133°C to 135°C
Quantity 10 g
Formula Weight 266.38
Percent Purity 95%
Chemical Name or Material 2',3',4',5',6'-Pentamethyl-2-phenylacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'