missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98% Product Code.: 15490547

Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98%

Product Code. 15490547
25 g
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy

Product Code. 15490547

Brand: Thermo Scientific Chemicals J66303.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose is used in the syntheses of ovalicin1 and of the sugar core of hikizimycin.

Solubility
Soluble in acetone, methanol, water.

Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14131-84-1
Molecular Formula C12H20O6
Molecular Weight (g/mol) 260.286
MDL Number MFCD00134206
InChI Key JWWCLCNPTZHVLF-WVHJSVDISA-N
Synonym 2,3:5,6-bis-o-1-methylethylidene-, a-d-mannofuranose, 3ar,4s,6r-6-2,2-dimethyl-1,3-dioxolan-4-yl-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-olShow MoreShow Less
PubChem CID 45039045
IUPAC Name (3aR,4S,6R)-6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
SMILES CC1(OCC(O1)C2C3C(C(O2)O)OC(O3)(C)C)C

Specifications

Melting Point 125°C to 126°C (decomposition)
Color White
Quantity 25 g
Beilstein 84382
Sensitivity Hygroscopic
Solubility Information Soluble in acetone,methanol,water.
Formula Weight 260.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3:5,6-Di-O-isopropylidene-alpha-D-mannofuranose

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM09E002 Certificate TypeCertificate of Analysis Date21/01/2025 Catalog NumberJ66303.09,J66303.06,J66303.14
Lot NumberM12K037 Certificate TypeCertificate of Analysis Date22/01/2024 Catalog NumberJ66303.09,J66303.06,J66303.14
Lot NumberX05D021 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ66303.09,J66303.06,J66303.14
Lot NumberU05A016 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ66303.09,J66303.06,J66303.14
Lot NumberM09E002 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberJ66303.09,J66303.06,J66303.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 2,3:4,6-Di-O-isopropylidene-alpha-L-sorbofuranose, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.