missing translation for 'onlineSavingsMsg'
Learn More

2-(3,4-Dimethoxyphenyl)ethanol, 98%, Thermo Scientific Chemicals

Product Code. 11451530
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7417-21-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.22
MDL Number MFCD00002894
InChI Key SRQAJMUHZROVHW-UHFFFAOYSA-N
Synonym 3,4-dimethoxyphenethyl alcohol, 2-3,4-dimethoxyphenyl ethanol, 3,4-dimethoxyphenethylalcohol, 2-3,4-dimethoxyphenyl ethan-1-ol, 3,4-dimethoxyphenethanol, homoveratryl alcohol, benzeneethanol, 3,4-dimethoxy, 3,4-dimethoxybenzeneethanol, 3,4-dimethoxy-.beta.-phenethyl alcohol, 1-2-hydroxyethyl-3,4-dimethoxybenzene
PubChem CID 81911
SMILES COC1=CC=C(CCO)C=C1OC

Specifications

Melting Point 45°C to 49°C
Boiling Point 172°C to 174°C (17 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
IUPAC Name 2-(3,4-dimethoxyphenyl)ethan-1-ol
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)ethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'