missing translation for 'onlineSavingsMsg'
Learn More

2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride, 95%, Thermo Scientific Chemicals

Product Code. 15455556
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1653-64-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035193
InChI Key NDYXFQODWGEGNU-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
PubChem CID 74246
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride
SMILES C1OC2=C(O1)C=C(C=C2)CCN.Cl

Specifications

Melting Point 213°C to 214°C
Quantity 1 g
Formula Weight 201.65
Percent Purity 95%
Chemical Name or Material 2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'