missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorophenol, 98%

Product Code. 10072580 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10072580

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 769-39-1 | C6H2F4O | 166.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-39-1
Molecular Formula C6H2F4O
Molecular Weight (g/mol) 166.07
MDL Number MFCD00002157
InChI Key PBYIIRLNRCVTMQ-UHFFFAOYSA-N
Synonym phenol, 2,3,5,6-tetrafluoro, unii-tna118i5tg, 2,3,5,6-tetrafluoro phenol, tna118i5tg, 2,3,5,6-tetrafluor-phenol, ccris 5843, zlchem 411, pubchem2135, acmc-1bc5w, 2,3,5,6tetrafluorophenol
PubChem CID 69858
IUPAC Name 2,3,5,6-tetrafluorophenol
SMILES C1=C(C(=C(C(=C1F)F)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 38°C
Color White
Boiling Point 140°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HC6F4OH
Quantity 5 g
Formula Weight 166.07
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2,3,5,6-Tetrafluorophenol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,5,6-Tetrafluorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.