missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tribromothiophene, 98%, Thermo Scientific™

Product Code. 11414120
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3141-24-0
Molecular Formula C4HBr3S
Molecular Weight (g/mol) 320.82
MDL Number MFCD00014521
InChI Key SKDNDSLDRLEELJ-UHFFFAOYSA-N
Synonym thiophene, 2,3,5-tribromo, pubchem5608, 2,5-tribromothiophene, acmc-209hmg, thiophene,3,5-tribromo, 2,3,5-tribromo thiophene, ksc225k9l, 2,3,5-tribromothiophene
PubChem CID 76592
IUPAC Name 2,3,5-tribromothiophene
SMILES BrC1=CC(Br)=C(Br)S1

Specifications

Melting Point 25°C to 29°C
Density 2.483
Boiling Point 77°C to 78°C (0.7 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.664
Quantity 100 g
UN Number UN2811
Beilstein 111486
Formula Weight 320.84
Percent Purity 98%
Chemical Name or Material 2,3,5-Tribromothiophene
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'