missing translation for 'onlineSavingsMsg'
Learn More

2',3',6'-Trifluoroacetophenone, 97%, Thermo Scientific™

Product Code. 11405766
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 208173-22-2
Molecular Formula C8H5F3O
Molecular Weight (g/mol) 174.122
MDL Number MFCD00070809
InChI Key INDGVPJYDRJJPJ-UHFFFAOYSA-N
Synonym 2',3',6'-trifluoroacetophenone, 1-2,3,6-trifluorophenyl ethanone, 2,3,6-trifluoroacetophenone, ethanone, 1-2,3,6-trifluorophenyl, 1-acetyl-2,3,6-trifluorobenzene, pubchem4285, acmc-1cnfl, 1-2,3,6-trifluorophenyl ethanone #
PubChem CID 594247
IUPAC Name 1-(2,3,6-trifluorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1F)F)F

Specifications

Refractive Index 1.467
Quantity 1 g
Formula Weight 174.12
Percent Purity 97%
Chemical Name or Material 2',3',6'-Trifluoroacetophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'