missing translation for 'onlineSavingsMsg'
Learn More

2-[3-(Chloromethyl)piperidino]-6-methylpyrazine, 97%, Thermo Scientific™

49.90 € - 119.00 €

Chemical Identifiers

CAS 937795-92-1
Molecular Formula C11H16ClN3
Molecular Weight (g/mol) 225.72
MDL Number MFCD09879918
InChI Key DTVYKEHYUMEUFH-UHFFFAOYNA-N
Synonym 2-3-chloromethyl piperidin-1-yl-6-methylpyrazine, 2-3-chloromethyl piperidino-6-methylpyrazine, 2-3-chloromethyl piperidyl-6-methylpyrazine
PubChem CID 24229601
IUPAC Name 2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
SMILES CC1=CN=CC(=N1)N1CCCC(CCl)C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10738265
View Documents Promotion Details
Thermo Scientific Maybridge
CC59024CB
250 mg Amber Glass Bottle
49.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10472474
View Documents Promotion Details
Thermo Scientific Maybridge
CC59024DA
1 g Amber Glass Bottle
119.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

937795-92-1
225.72
DTVYKEHYUMEUFH-UHFFFAOYNA-N
24229601
CC1=CN=CC(=N1)N1CCCC(CCl)C1
C11H16ClN3
MFCD09879918
2-3-chloromethyl piperidin-1-yl-6-methylpyrazine, 2-3-chloromethyl piperidino-6-methylpyrazine, 2-3-chloromethyl piperidyl-6-methylpyrazine
2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine

Specifications

937795-92-1
100.0
C11H16ClN3
2-3-chloromethyl piperidin-1-yl-6-methylpyrazine, 2-3-chloromethyl piperidino-6-methylpyrazine, 2-3-chloromethyl piperidyl-6-methylpyrazine
CC1=CN=CC(=N1)N1CCCC(CCl)C1
225.72
225.72
2-[3-(Chloromethyl)piperidino]-6-methylpyrazine
96.5
97%
MFCD09879918
DTVYKEHYUMEUFH-UHFFFAOYNA-N
2-[3-(chloromethyl)piperidin-1-yl]-6-methylpyrazine
24229601
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications