missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-1-propanol, 97%, Thermo Scientific Chemicals

Product Code. 10669543
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-23-9
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.99
MDL Number MFCD00040446
InChI Key ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
Synonym 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
PubChem CID 12018
IUPAC Name 2,3-dichloropropan-1-ol
SMILES C(C(CCl)Cl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.3500g/mL
Boiling Point 182°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 5 g
Specific Gravity 1.35
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,acetone,ethanol
Formula Weight 128.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-1-propanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,3-Dichloro-1-propanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,3-Dichloro-1-propanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'