missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-5,6-dicyano-1,4-benzoquinone, 98%, Thermo Scientific Chemicals

Product Code. 11915611
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-58-2
Molecular Formula C8Cl2N2O2
Molecular Weight (g/mol) 227.01
InChI Key HZNVUJQVZSTENZ-UHFFFAOYSA-N
Synonym 2,3-dichloro-5,6-dicyano-1,4-benzoquinone, dichlorodicyanoquinone, dichlorodicyanobenzoquinone, dichlorodicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyano-p-benzoquinone, 2,3-dichloro-5,6-dicyanobenzoquinone, 1,4-cyclohexadiene-1,2-dicarbonitrile, 4,5-dichloro-3,6-dioxo, ddq, 2,3-dichloro-5,6-dicyanoquinone, 4,5-dichloro-3,6-dioxo-1,4-cyclohexadiene-1,2-dicarbonitrile
PubChem CID 6775
IUPAC Name 4,5-dichloro-3,6-dioxocyclohexa-1,4-diene-1,2-dicarbonitrile
SMILES C(#N)C1=C(C(=O)C(=C(C1=O)Cl)Cl)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 210.0°C to 217.0°C
Color Yellow to Orange
Loss on Drying 0.5% max. (1 g, 80°C, 2 hrs, 40mm Hg vacuum)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Iodometry)
Packaging Glass bottle
Quantity 500 g
Beilstein 10,902
Fieser 01,215; 02,112; 03,83; 04,130; 05,193; 06,168; 07,96; 08,153; 09,148; 10,135; 11,166; 13,104; 14,126; 15,125; 16,120
Merck Index 15,3072
Solubility Information Solubility in water: reacts. Other solubilities: soluble in benzene,dioxane,acetic acid,ethanol,slightly soluble in chloroform,methylenechloride
Formula Weight 227.01
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,3-Dichloro-5,6-dicyano-1,4-benzoquinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'