missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloro-N'-hydroxybenzenecarboximidamide, 97%, Thermo Scientific™

Product Code. 10497822
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 261761-55-1
Molecular Formula C7H6Cl2N2O
Molecular Weight (g/mol) 205.038
MDL Number MFCD01567248
InChI Key OPOPCNCEDLBYPV-UHFFFAOYSA-N
Synonym 2,3-dichlorobenzamidoxime, hcmhpiwpbbblbbbftocadcpba`, 2,3-dichlorophenyl hydroxyimino methylamine, z-2,3-dichloro-n'-hydroxybenzene-1-carboximidamide
PubChem CID 9580371
IUPAC Name 2,3-dichloro-N'-hydroxybenzenecarboximidamide
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3C(=NOH)NH2
Quantity 250 mg
Formula Weight 205.04
Percent Purity 97%
Chemical Name or Material 2, 3-Dichloro-N'-hydroxybenzenecarboximidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.