missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorobenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11498783
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-45-3
Molecular Formula C7H4Cl2O2
Molecular Weight (g/mol) 191.007
MDL Number MFCD00002413
InChI Key QAOJBHRZQQDFHA-UHFFFAOYSA-N
Synonym benzoic acid, 2,3-dichloro, 2,3-dichloro benzoic acid, unii-cgp39qv5wf, benzoic acid, dichloro, 2,3-dichorobenzoic acid, cgp39qv5wf, dichlorobenzoic acid, lamotrigine impurity e, pubchem8198, acmc-209kmq
PubChem CID 5765
IUPAC Name 2,3-dichlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)Cl)C(=O)O

Specifications

Melting Point 166°C to 170°C
Quantity 10 g
Beilstein 1946217
Formula Weight 191.01
Percent Purity 98%
Chemical Name or Material 2,3-Dichlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'