missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorooctafluorobutane, 97%, Thermo Scientific™

21.31 € - 69.20 €

Chemical Identifiers

CAS 355-20-4
Molecular Formula C4Cl2F8
Molecular Weight (g/mol) 270.93
MDL Number MFCD00018064
InChI Key LXANZHXWGZWFAC-UHFFFAOYNA-N
PubChem CID 78982
IUPAC Name 2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
SMILES FC(F)(F)C(F)(Cl)C(F)(Cl)C(F)(F)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307316
View Documents
Thermo Scientific Alfa Aesar
L09252.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11317316
View Documents
Thermo Scientific Alfa Aesar
L09252.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

355-20-4
270.93
LXANZHXWGZWFAC-UHFFFAOYNA-N
2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
C4Cl2F8
MFCD00018064
78982
FC(F)(F)C(F)(Cl)C(F)(Cl)C(F)(F)F

Specifications

355-20-4
1.68
C4Cl2F8
MFCD00018064
2,3-dichlorooctafluorobutane, butane, 2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro, butane, 2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro-, r*,r*, acmc-1cpcx, acmc-20dh57, 2,3-dichlorooctafluorobutan, butane,2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro-, 2r,3r-rel, #, butane,2,3-dichloro-1,1,1,2,3,4,4,4-octafluoro
FC(F)(F)C(F)(Cl)C(F)(Cl)C(F)(F)F
270.93
270.94
2,3-Dichlorooctafluorobutane
∼-68°C
62°C to 63°C
1.313
1777520
LXANZHXWGZWFAC-UHFFFAOYNA-N
2,3-dichloro-1,1,1,2,3,4,4,4-octafluorobutane
78982
97%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 206-578-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.