missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichlorophenol, 98+%, Thermo Scientific Chemicals

Product Code. 11420018
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-24-9
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002160
InChI Key UMPSXRYVXUPCOS-UHFFFAOYSA-N
Synonym phenol, 2,3-dichloro, dichlorophenol, unii-izm39u444l, ccris 5902, 2,3-dichloro phenol, 2,3 dichlorophenol, phenol,3-dichloro, pubchem3696, 2,3-dichloro-phenol, acmc-1ar0l
PubChem CID 11334
IUPAC Name 2,3-dichlorophenol
SMILES OC1=CC=CC(Cl)=C1Cl

Specifications

Melting Point 57°C to 60°C
Boiling Point 206°C
Flash Point 115°C (239°F)
Odor Phenol-like
Quantity 100 g
UN Number UN2020
Beilstein 2043615
Formula Weight 163
Percent Purity ≥98%
Chemical Name or Material 2,3-Dichlorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'