Learn More
2,3-Dichlorophenylacetic acid, 97%, Thermo Scientific™
40.10 € - 130.65 €
Chemical Identifiers
CAS | 10236-60-9 |
---|---|
Molecular Formula | C8H6Cl2O2 |
Molecular Weight (g/mol) | 205.04 |
MDL Number | MFCD01861393 |
InChI Key | YWMXEUIQZOQESD-UHFFFAOYSA-N |
Synonym | 2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid |
PubChem CID | 2734600 |
IUPAC Name | 2-(2,3-dichlorophenyl)acetic acid |
SMILES | C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10334452
|
Thermo Scientific
365550050 |
5 g | Glass bottle |
40.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10570944
|
Thermo Scientific Acros
365550250 |
25 g | Glass bottle |
130.65 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
10236-60-9 | |
205.04 | |
YWMXEUIQZOQESD-UHFFFAOYSA-N | |
2734600 | |
C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O |
C8H6Cl2O2 | |
MFCD01861393 | |
2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid | |
2-(2,3-dichlorophenyl)acetic acid |
Specifications
10236-60-9 | |
100.0 | |
White | |
96% min. (HPLC) | |
Cl2C6H3CH2COOH | |
2,3-dichlorophenylacetic acid, 2-2,3-dichlorophenyl acetic acid, benzeneacetic acid, 2,3-dichloro, 2,3-dichlorophenyl acetic acid, 2,3-dichlorobenzeneacetic acid, rarechem al bo 1228, timtec-bb sbb003500, 3-dichlorophenylacetic acid, acmc-1c25x, 2,3-dichlorophenylaceticacid | |
C1=CC(=C(C(=C1)Cl)Cl)CC(=O)O | |
205.04 | |
205.04 | |
Powder |
96.0 | |
130°C to 135°C | |
Authentic | |
C8H6Cl2O2 | |
MFCD01861393 | |
YWMXEUIQZOQESD-UHFFFAOYSA-N | |
2-(2,3-dichlorophenyl)acetic acid | |
2734600 | |
97% | |
2, 3-Dichlorophenylacetic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
- 2,3-Dichlorophenylacetic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.