missing translation for 'onlineSavingsMsg'
Learn More

2,3-Dichloropyrazine, 98%, Thermo Scientific Chemicals

94.97 € - 1318.56 €

Chemical Identifiers

CAS 4858-85-9
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00040964
InChI Key MLCNOCRGSBCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
PubChem CID 78575
IUPAC Name 2,3-dichloropyrazine
SMILES ClC1=NC=CN=C1Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10680735
View Documents
Thermo Scientific Acros
377040050
5 g
94.97 €
5g
Please to purchase this item. Need a web account? Register with us today!
10591774
View Documents
Thermo Scientific Acros
377040250
25 g
330.65 €
25g
Please to purchase this item. Need a web account? Register with us today!
11946631
View Documents
Thermo Scientific Acros
377041000
100 g
1318.56 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4858-85-9
148.97
MLCNOCRGSBCAGH-UHFFFAOYSA-N
78575
ClC1=NC=CN=C1Cl
C4H2Cl2N2
MFCD00040964
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
2,3-dichloropyrazine

Specifications

4858-85-9
100.0
1.4310g/mL
102°C
Glass bottle
MFCD00040964
pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
MLCNOCRGSBCAGH-UHFFFAOYSA-N
2,3-dichloropyrazine
78575
98%
97.5
22.5°C to 24.5°C
105.0°C to 107.0°C (50.0 mmHg)
97.5% min. (GC)
C4H2Cl2N2
1.431
Solubility in water: insoluble
ClC1=NC=CN=C1Cl
148.97
148.98
2, 3-Dichloropyrazine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 225-460-1

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.